Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other STYXL1 SIRNA Assays ›
Gene Symbol:
STYXL1
TMEM120A
Gene Name:
serine/threonine/tyrosine interacting-like 1
transmembrane protein 120A
Gene Aliases:
DUSP24, MK-STYX, MKSTYX
NET29, TMPIT
Chromosome Location:
Chr.7: 75995820 - 76048709 on Build GRCh38
Chromosome Location:
Chr.7: 75986837 - 75994674 on Build GRCh38
UniGene ID:
Hs.11615
Hs.488835
Species:
Human
siRNA ID
HSS122486
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: STYXL1
Interrogated Sequence
Translated Protein
RNADB ID
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001317785.1
NP_001304714.1
9
1235
NM_001317786.1
NP_001304715.1
9
1085
NM_001317787.1
NP_001304716.1
7
794
NM_001317788.1
NP_001304717.1
7
747
NM_001317789.1
NP_001304718.1
7
744
NM_016086.2
NP_057170.1
9
1232
NR_134486.1
7
823
NR_134487.1
7
777
XM_011516292.2
XP_011514594.1
8
965
XM_011516293.2
XP_011514595.1
9
1078
XM_011516300.1
XP_011514602.1
8
888
XM_017012296.1
XP_016867785.1
10
1192
XM_017012297.1
XP_016867786.1
8
968
XM_017012298.1
XP_016867787.1
8
928
XM_017012299.1
XP_016867788.1
10
1145
XM_017012300.1
XP_016867789.1
10
1112
XM_017012301.1
XP_016867790.1
8
932
XM_017012302.1
XP_016867791.1
8
935
GenBank mRNA
AA782050.1
-
1
185
AF069762.1
-
9
1228
AF188201.1
-
6
641
AF188202.1
-
5
466
AF188203.1
-
5
496
AF188204.1
-
6
609
AI676056.1
-
1
181
AY927581.1
-
LIT3580
AY927581
9
989
BC024035.2
-
9
1195
2988900
BM972220.1
-
1
198
BX537896.1
-
7
774
W58723.1
-
3
280
Gene Symbol: TMEM120A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
BI049925.1
-
2
191
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
intracellular signal transduction
protein tyrosine/serine/threonine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline