Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 10 | 1871 | ||
GenBank mRNA | AJ314790.1 | - | 10 | 1824 | ||
AK292214.1 | - | 10 | 1871 | |||
BC030540.1 | - | 10 | 1843 | |||
BC051893.1 | - | 3 | 462 | |||
BC062986.1 | - | 3 | 387 | |||
BC137084.1 | - | 10 | 1835 | 9021081 | ||
BC137085.1 | - | 10 | 1835 | 9021082 |