Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 4 | 615 | ||
GenBank mRNA | AK292214.1 | - | 4 | 615 | ||
AK309638.1 | - | 4 | 615 | |||
BC030540.1 | - | 4 | 587 | |||
BC107417.1 | - | 4 | 566 | |||
BC137084.1 | - | 4 | 579 | 9021081 | ||
BC137085.1 | - | 4 | 579 | 9021082 |