Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016116.2 | NP_057200.1 | 1 | 226 | ||
NM_145872.2 | NP_665879.1 | 1 | 226 | |||
XM_017012303.1 | XP_016867792.1 | 1 | 688 | |||
GenBank mRNA | AF156779.1 | - | 1 | 155 | ||
BC025024.1 | - | 1 | 64 | |||
BC113479.1 | - | 1 | 226 | 8322531 | ||
BC113481.1 | - | 1 | 226 | 8322533 |