Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUFU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178133.1 | NP_001171604.1 | 9 | 1292 | ||
NM_016169.3 | NP_057253.2 | 9 | 1292 | |||
XM_011539858.2 | XP_011538160.1 | 9 | 1295 | |||
XM_011539860.2 | XP_011538162.1 | 9 | 1292 | |||
XM_011539861.2 | XP_011538163.1 | 9 | 1295 | |||
XM_011539863.2 | XP_011538165.1 | 9 | 1030 | |||
XM_011539864.2 | XP_011538166.1 | 9 | 1295 | |||
XM_017016323.1 | XP_016871812.1 | 9 | 1525 | |||
GenBank mRNA | AF144231.1 | - | 9 | 1247 | ||
AF159447.1 | - | 9 | 1247 | |||
AF172319.1 | - | 9 | 1097 | |||
AF175770.1 | - | 9 | 1267 | |||
AF222345.1 | - | 9 | 1175 | |||
AK307370.1 | - | 4 | 501 | |||
AK311503.1 | - | 4 | 470 | |||
AK312319.1 | - | 9 | 1282 | |||
AY358550.1 | - | 9 | 1247 | |||
BC013291.2 | - | 9 | 1268 | 3533158 |