Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318524.1 | NP_001305453.1 | 2, 3 | 266 | ||
NM_001318525.1 | NP_001305454.1 | 2, 3 | 266 | |||
NM_001318528.1 | NP_001305457.1 | 2, 3 | 243 | |||
NM_001318530.1 | NP_001305459.1 | 2, 3 | 243 | |||
NM_016209.4 | NP_057293.1 | 2, 3 | 266 | |||
GenBank mRNA | AF089106.1 | - | 2, 3 | 259 | ||
AF161524.1 | - | 2, 3 | 221 | |||
AK126779.1 | - | 2, 3 | 225 | |||
AK311885.1 | - | 2, 3 | 235 | |||
AX771012.1 | - | 2, 3 | 259 | |||
AX771013.1 | - | 2, 3 | 221 | |||
BC011369.1 | - | 2, 3 | 213 | 3868798 | ||
BC018024.1 | - | 2, 3 | 220 | 4287920 | ||
BC105809.1 | - | 2, 3 | 232 | 6745065 | ||
BX419849.2 | - | 2, 3 | 211 | |||
DA209193.1 | - | 2, 3 | 243 | |||
DA293122.1 | - | 2, 3 | 266 | |||
DA295904.1 | - | 2, 3 | 202 | |||
DA833474.1 | - | 2, 3 | 205 |