Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282150.1 | NP_001269079.1 | 4, 5 | 435 | ||
NM_016226.4 | NP_057310.1 | 2, 3 | 280 | |||
NM_057180.2 | NP_476528.1 | 3, 4 | 292 | |||
NR_104099.1 | 4, 5 | 332 | ||||
NR_104100.1 | 3, 4 | 320 | ||||
NR_104101.1 | 3, 4 | 370 | ||||
NR_104102.1 | 2, 3 | 171 | ||||
XM_006719459.3 | XP_006719522.1 | 4, 5 | 341 | |||
XM_006719460.3 | XP_006719523.1 | 5, 6 | 378 | |||
XM_011538484.2 | XP_011536786.1 | 3, 4 | 326 | |||
XM_017019457.1 | XP_016874946.1 | 3, 4 | 351 | |||
XM_017019458.1 | XP_016874947.1 | 2, 3 | 471 | |||
GenBank mRNA | AA148059.1 | - | 2, 3 | 118 | ||
AB578890.1 | - | 1, 2 | 19 | |||
AF168716.1 | - | 3, 4 | 250 | |||
AF175264.1 | - | 2, 3 | 249 | |||
AF193795.1 | - | 2, 3 | 242 | |||
AF201936.1 | - | 3, 4 | 337 | |||
AF201946.1 | - | 2, 3 | 228 | |||
AK129767.1 | - | 2, 3 | 242 | |||
AK130510.1 | - | 1078 | ||||
AL136614.1 | - | 2, 3 | 258 | |||
AL832866.1 | - | 2, 3 | 2273 | |||
BC000880.1 | - | 3, 4 | 250 | 3461977 | ||
BC015095.1 | - | 3, 4 | 259 | |||
BC095446.1 | - | 2, 3 | 251 | 30719438 | ||
BF030144.1 | - | 291 | ||||
BG707325.1 | - | 2, 3 | 275 | |||
BM806646.1 | - | 4, 5 | 388 | |||
BU568689.1 | - | 4, 5 | 293 | |||
CR457182.1 | - | 1, 2 | 203 | |||
CR533468.1 | - | 1, 2 | 191 |