Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016332.2 | NP_057416.1 | 3, 4 | 447 | ||
GenBank mRNA | AF161388.1 | - | 3, 4 | 404 | ||
AF166124.1 | - | 3, 4 | 406 | |||
AF187272.1 | - | 3, 4 | 381 | |||
AK026128.1 | - | 3, 4 | 418 | |||
BC003127.1 | - | 3, 4 | 343 | 2905838 |