Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166663.1 | NP_001160135.1 | 2 | 445 | ||
NM_001166664.1 | NP_001160136.1 | 2 | 445 | |||
NM_016382.3 | NP_057466.1 | 2 | 445 | |||
XM_011509621.2 | XP_011507923.1 | 2 | 430 | |||
XM_011509622.2 | XP_011507924.1 | 2 | 430 | |||
GenBank mRNA | AF105261.1 | - | 2 | 262 | ||
AF107761.2 | - | 2 | 328 | |||
AF117711.1 | - | 2 | 399 | |||
AF145782.1 | - | 2 | 220 | |||
AF242540.1 | - | 2 | 303 | |||
AF363452.1 | - | 1 | 141 | |||
AJ245375.1 | - | 2 | 243 | |||
AJ245376.1 | - | 2 | 243 | |||
AJ245377.1 | - | 2 | 243 | |||
AK295950.1 | - | 2 | 430 | |||
AK313201.1 | - | 2 | 366 | |||
BC028073.1 | - | 2 | 445 | 5213370 | ||
BC041607.1 | - | 2 | 247 | |||
BC053985.1 | - | 2 | 231 | 5484877 |