Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM8B SIRNA Assays ›
Gene Symbol:
TMEM8B
Gene Name:
transmembrane protein 8B
Gene Aliases:
C9orf127, NAG-5, NAG5, NGX6
Chromosome Location:
Chr.9: 35825817 - 35865515 on Build GRCh38
UniGene ID:
Hs.493808
Species:
Human
siRNA ID
HSS122609
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM8B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042589.2
NP_001036054.1
11
1816
NM_001042590.2
NP_001036055.1
10
2100
NM_016446.3
NP_057530.2
11
1888
XM_005251483.4
XP_005251540.3
10
2070
XM_011517901.2
XP_011516203.1
11
2148
XM_011517902.2
XP_011516204.1
11
2148
XM_011517903.2
XP_011516205.1
11
2148
XM_011517904.2
XP_011516206.1
10
2271
XM_011517905.1
XP_011516207.1
11
2092
XM_011517908.2
XP_011516210.1
10
1601
XM_011517910.2
XP_011516212.1
10
1479
XM_011517911.2
XP_011516213.1
12
1894
XM_011517912.2
XP_011516214.1
11
1740
XM_011517913.2
XP_011516215.1
11
1955
XM_011517914.2
XP_011516216.1
9
1400
XM_011517915.2
XP_011516217.1
9
1699
XM_011517916.2
XP_011516218.1
6
783
XM_011517918.2
XP_011516220.1
6
601
XM_017014805.1
XP_016870294.1
9
1539
XM_017014806.1
XP_016870295.1
9
1513
XM_017014807.1
XP_016870296.1
5
732
GenBank mRNA
AF070572.1
-
9
1537
AF188239.1
-
11
1849
AK074844.1
-
9
2211
BC082765.1
-
10
2013
BC098265.1
-
8
1431
40025067
BC098296.1
-
8
1431
40025068
BC099733.1
-
8
1431
40025069
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
regulation of mitotic cell cycle
regulation of growth
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0