Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016520.2 | NP_057604.1 | 7 | 721 | ||
XM_011518780.2 | XP_011517082.1 | 6 | 658 | |||
GenBank mRNA | AF151054.1 | - | 7 | 712 | ||
AF218421.1 | - | 7 | 676 | |||
AK057004.1 | - | 7 | 703 | |||
AK096971.1 | - | 1 | 1251 | |||
AK097577.1 | - | 4 | 1483 | |||
AL137549.1 | - | 5 | 507 | |||
AY632862.1 | - | 4 | 412 | |||
BC007664.2 | - | 7 | 674 | 3140172 | ||
BC017570.1 | - | 7 | 678 | 4109447 | ||
BC037173.1 | - | 2 | 227 |