Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278603.1 | NP_001265532.1 | 9 | 1249 | ||
NM_017426.3 | NP_059122.2 | 10 | 1393 | |||
NR_103780.1 | 9 | 1166 | ||||
NR_103781.1 | 8 | 1058 | ||||
NR_103782.1 | 10 | 1163 | ||||
XM_006714237.1 | XP_006714300.1 | 11 | 1370 | |||
XM_011532033.2 | XP_011530335.1 | 9 | 1228 | |||
XM_011532034.2 | XP_011530336.1 | 9 | 1636 | |||
XM_017008294.1 | XP_016863783.1 | 10 | 1780 | |||
XM_017008295.1 | XP_016863784.1 | 9 | 1553 | |||
GenBank mRNA | AF157322.1 | - | 10 | 1271 | ||
AK001517.1 | - | 10 | 1275 | |||
AK222752.1 | - | 10 | 1280 | |||
AK223352.1 | - | 10 | 1276 | |||
AK299442.1 | - | 9 | 1048 | |||
AK300036.1 | - | 9 | 1131 | |||
AK315160.1 | - | 10 | 1273 | |||
AL137650.1 | - | 3 | 392 | |||
BC012559.1 | - | 10 | 1290 | |||
BX538002.1 | - | 8 | 958 |