Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016930.2 | NP_058626.1 | 1, 2 | 250 | ||
XM_011513480.1 | XP_011511782.1 | 1, 2 | 229 | |||
XM_011513483.1 | XP_011511785.1 | 1, 2 | 229 | |||
XM_017008296.1 | XP_016863785.1 | 1, 2 | 229 | |||
XM_017008298.1 | XP_016863787.1 | 1, 2 | 229 | |||
GenBank mRNA | AB028741.1 | - | 1, 2 | 169 | ||
AK310026.1 | - | 1, 2 | 212 | |||
AY453396.1 | - | 1, 2 | 166 | |||
BC014613.2 | - | 1, 2 | 172 | 2960768 | ||
BG771856.1 | - | 1, 2 | 255 | |||
BT007150.1 | - | 1, 2 | 166 | |||
BT020135.1 | - | 1, 2 | 166 |