Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016930.2 | NP_058626.1 | 7 | 718 | ||
XM_011513480.1 | XP_011511782.1 | 8 | 790 | |||
XM_011513483.1 | XP_011511785.1 | 8 | 750 | |||
XM_011513485.2 | XP_011511787.1 | 7 | 645 | |||
XM_017008297.1 | XP_016863786.1 | 8 | 1371 | |||
GenBank mRNA | AB028741.1 | - | 7 | 637 | ||
AK310026.1 | - | 7 | 680 | |||
AY453396.1 | - | 7 | 634 | |||
BC014613.2 | - | 7 | 640 | 2960768 | ||
BT007150.1 | - | 7 | 634 | |||
BT020135.1 | - | 7 | 634 |