Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017429.2 | NP_059125.2 | 10 | 1543 | ||
XM_011523109.1 | XP_011521411.1 | 8 | 1391 | |||
XM_017023288.1 | XP_016878777.1 | 9 | 1497 | |||
XM_017023289.1 | XP_016878778.1 | 6 | 657 | |||
GenBank mRNA | AF294900.1 | - | 10 | 1543 | ||
AK001592.1 | - | 10 | 1543 | |||
AK296011.1 | - | 9 | 1415 | |||
BC126210.1 | - | 10 | 1445 | 8991926 | ||
BC126212.1 | - | 10 | 1445 | 8991928 |