Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015946.4 | NP_057030.3 | 2 | 1507 | ||
GenBank mRNA | AF132951.1 | - | 1 | 781 | ||
AF139828.1 | - | 1 | 754 | |||
AK025729.1 | - | 1 | 821 | |||
AK056144.1 | - | 2 | 1507 | |||
AK312633.1 | - | 1 | 759 | |||
AY117399.1 | - | 2 | 1507 | |||
BC005889.2 | - | 1 | 795 | 2967612 | ||
BC007249.2 | - | 1 | 795 | 2967002 | ||
BC007650.1 | - | 1 | 812 | 3608639 | ||
BC022789.1 | - | 1 | 706 | 3160699 |