Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145944.1 | NP_001139416.1 | 2, 3 | 481 | ||
NM_001145945.1 | NP_001139417.1 | 5, 6 | 764 | |||
NM_001145946.1 | NP_001139418.1 | 5, 6 | 764 | |||
NM_017435.4 | NP_059131.1 | 4, 5 | 777 | |||
XM_005253394.2 | XP_005253451.1 | 5, 6 | 553 | |||
XM_005253396.2 | XP_005253453.1 | 3, 4 | 5318 | |||
XM_011520703.2 | XP_011519005.1 | 5, 6 | 553 | |||
XM_011520704.2 | XP_011519006.1 | 5, 6 | 553 | |||
XM_011520709.2 | XP_011519011.1 | 2, 3 | 195 | |||
XM_017019484.1 | XP_016874973.1 | 7, 8 | 5814 | |||
XM_017019485.1 | XP_016874974.1 | 6, 7 | 5732 | |||
XM_017019486.1 | XP_016874975.1 | 4, 5 | 5472 | |||
XM_017019487.1 | XP_016874976.1 | 3, 4 | 5318 | |||
GenBank mRNA | AF205076.1 | - | 4, 5 | 778 | ||
AF260704.1 | - | 4, 5 | 473 | |||
AK294333.1 | - | 2, 3 | 349 | |||
AK296236.1 | - | 5, 6 | 657 | |||
AK303713.1 | - | 5, 6 | 640 | |||
AL834209.1 | - | 5, 6 | 629 | |||
BC022461.2 | - | 4, 5 | 506 | 4801171 |