Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243270.1 | NP_001230199.1 | 10, 11 | 1669 | ||
NM_001243271.1 | NP_001230200.1 | 11, 12 | 1808 | |||
NM_014361.3 | NP_055176.1 | 11, 12 | 1808 | |||
NM_175566.2 | NP_780775.1 | 8, 9 | 1152 | |||
XM_011542871.1 | XP_011541173.1 | 10, 11 | 1586 | |||
XM_011542873.1 | XP_011541175.1 | 11, 12 | 1808 | |||
XM_017017926.1 | XP_016873415.1 | 12, 13 | 1976 | |||
XM_017017927.1 | XP_016873416.1 | 12, 13 | 1976 | |||
XM_017017928.1 | XP_016873417.1 | 10, 11 | 1741 | |||
XM_017017929.1 | XP_016873418.1 | 10, 11 | 1586 | |||
GenBank mRNA | AB013802.2 | - | 9, 10 | 1383 | ||
AB013803.2 | - | 8, 9 | 1161 | |||
AK308788.1 | - | 11, 12 | 1808 | |||
BC039255.1 | - | 11, 12 | 1643 | 5273769 | ||
BC130619.1 | - | 10, 11 | 1455 | 40146650 | ||
BC144185.1 | - | 10, 11 | 1455 | 9052705 | ||
BC144186.1 | - | 11, 12 | 1594 | 9052706 |