Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322270.1 | NP_001309199.1 | 13 | 2010 | ||
NM_001322271.1 | NP_001309200.1 | 13 | 2003 | |||
NM_001322272.1 | NP_001309201.1 | 14 | 2368 | |||
NM_017437.2 | NP_059133.1 | 14 | 2163 | |||
GenBank mRNA | AB037788.1 | - | 8 | 1287 | ||
AK001627.2 | - | 14 | 2140 | |||
AK001912.1 | - | 12 | 1719 | |||
AK023583.1 | - | 11 | 1740 | |||
AL442079.1 | - | 7 | 1062 | |||
BC070095.1 | - | 14 | 2133 | 5267724 |