Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPEAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272037.1 | NP_001258966.1 | 12 | 1961 | ||
NM_144991.2 | NP_659428.2 | 11 | 1899 | |||
GenBank mRNA | AJ487962.1 | - | 11 | 1899 | ||
BC115415.1 | - | 9 | 1425 | 40080529 |