Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017446.3 | NP_059142.2 | 2 | 203 | ||
NM_080794.3 | NP_542984.2 | 2 | 203 | |||
XM_006724026.3 | XP_006724089.1 | 2 | 195 | |||
XM_011529651.2 | XP_011527953.1 | 2 | 274 | |||
GenBank mRNA | AF109357.1 | - | 2 | 177 | ||
AF239727.1 | - | 2 | 189 | |||
AK000458.1 | - | 2 | 195 | |||
AK225878.1 | - | 2 | 195 | |||
BC004896.2 | - | 2 | 176 | |||
BC107719.1 | - | 2 | 199 | 6737868 | ||
BE865858.1 | - | 202 | ||||
BQ719968.1 | - | 2 | 231 |