Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308101.1 | NP_001295030.1 | 2 | 325 | ||
NM_020640.3 | NP_065691.2 | 2 | 307 | |||
XM_011512912.1 | XP_011511214.1 | 2 | 276 | |||
XM_011512914.1 | XP_011511216.1 | 2 | 315 | |||
XM_011512915.2 | XP_011511217.1 | 3 | 837 | |||
XM_011512916.2 | XP_011511218.1 | 2 | 765 | |||
XM_017006633.1 | XP_016862122.1 | 1 | 192 | |||
XM_017006634.1 | XP_016862123.1 | 2 | 281 | |||
XM_017006635.1 | XP_016862124.1 | 3 | 837 | |||
GenBank mRNA | AF292100.2 | - | 2 | 224 | ||
AF456425.1 | - | 2 | 210 | |||
AF456426.1 | - | 2 | 210 | |||
AK056335.1 | - | 2 | 240 | |||
AK297328.1 | - | 2 | 244 | |||
AK314480.1 | - | 2 | 224 | |||
BC009478.2 | - | 2 | 229 | 3939758 | ||
BC013163.1 | - | 1 | 180 | |||
BI828039.1 | - | 2 | 265 | |||
CD687300.1 | - | 2 | 285 | |||
HY031019.1 | - | 2 | 307 | |||
HY052047.1 | - | 2 | 322 |