Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERRFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018948.3 | NP_061821.1 | 4 | 1166 | ||
XM_005263477.2 | XP_005263534.1 | 4 | 1180 | |||
XM_006710697.2 | XP_006710760.1 | 4 | 2350 | |||
XM_011541596.2 | XP_011539898.1 | 3 | 1087 | |||
GenBank mRNA | AJ276373.1 | - | 3 | 917 | ||
AK027830.1 | - | 1 | 261 | |||
AK315718.1 | - | 4 | 1121 | |||
AL137274.1 | - | 1 | 15 | |||
BC025337.1 | - | 4 | 1129 | 4810371 |