Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146265.1 | NP_001139737.1 | 3 | 897 | ||
NM_001146266.1 | NP_001139738.1 | 2 | 787 | |||
NM_001146267.1 | NP_001139739.1 | 3 | 724 | |||
NM_018970.6 | NP_061843.3 | 3 | 894 | |||
XM_005250451.2 | XP_005250508.1 | 2 | 844 | |||
XM_017012352.1 | XP_016867841.1 | 3 | 959 | |||
GenBank mRNA | AB040800.1 | - | 1 | 270 | ||
AF250237.2 | - | 3 | 882 | |||
AK312587.1 | - | 2 | 662 | |||
AL161959.1 | - | 3 | 727 | |||
BC030577.1 | - | 3 | 848 | 4823671 | ||
DA483702.1 | - | 2 | 478 | |||
DA596250.1 | - | 1 | 420 |
Set Membership: |
G-protein coupled receptor |