Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040875.2 | NP_001035808.1 | 3 | 277 | ||
NM_018972.2 | NP_061845.2 | 3 | 407 | |||
NR_046346.1 | 3 | 262 | ||||
XM_011517552.2 | XP_011515854.1 | 2 | 202 | |||
XM_017013585.1 | XP_016869074.1 | 3 | 1539 | |||
XM_017013586.1 | XP_016869075.1 | 3 | 1539 | |||
GenBank mRNA | AB551556.1 | - | 3 | 406 | ||
AB551557.1 | - | 3 | 406 | |||
AB551558.1 | - | 3 | 404 | |||
AK292572.1 | - | 3 | 346 | |||
AK295594.1 | - | 3 | 201 | |||
BC024939.1 | - | 3 | 242 | 3923335 | ||
BC035450.1 | - | 3 | 254 | |||
DB505741.1 | - | 3 | 407 | |||
Y17849.1 | - | 3 | 335 |