Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271581.1 | NP_001258510.1 | 15 | 1954 | ||
NM_018981.2 | NP_061854.1 | 16 | 2092 | |||
NR_073365.1 | 16 | 2846 | ||||
NR_073366.1 | 16 | 2846 | ||||
NR_073367.1 | 14 | 1910 | ||||
GenBank mRNA | AF038503.1 | - | 16 | 1803 | ||
AF490904.1 | - | 15 | 1702 | |||
AK027450.1 | - | 14 | 1433 | |||
AK027647.1 | - | 16 | 1861 | |||
AK027696.1 | - | 16 | 1840 | |||
AK027701.2 | - | 13 | 1214 | |||
AK074905.1 | - | 3 | 208 | |||
AK298138.1 | - | 11 | 939 | |||
AL137648.1 | - | 5 | 642 | |||
AL832267.1 | - | 9 | 792 | |||
AL832632.2 | - | 16 | 2617 | |||
AL832646.3 | - | 14 | 1443 | |||
AY089971.1 | - | 12 | 1264 | |||
AY358577.1 | - | 15 | 1711 | |||
BC070034.1 | - | 16 | 2586 | |||
BC117299.1 | - | 16 | 1807 | 40125850 | ||
BC126168.1 | - | 16 | 1807 | 8991884 |