Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282857.1 | NP_001269786.1 | 36 | 4266 | ||
NM_019001.4 | NP_061874.3 | 36 | 4263 | |||
XM_011512919.2 | XP_011511221.1 | 36 | 4266 | |||
XM_011512920.2 | XP_011511222.1 | 36 | 4266 | |||
XM_011512922.2 | XP_011511224.1 | 33 | 3897 | |||
XM_017006640.1 | XP_016862129.1 | 36 | 4263 | |||
XM_017006641.1 | XP_016862130.1 | 36 | 4266 | |||
GenBank mRNA | AL133623.1 | - | 22 | 2544 | ||
AY137776.1 | - | 36 | 4145 | |||
BX640905.1 | - | 36 | 4212 | |||
CR627396.1 | - | 36 | 4218 | |||
CR749518.1 | - | 36 | 4196 |