Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009584.1 | NP_001009584.1 | 2 | 432 | ||
NM_001184768.1 | NP_001171697.1 | 4 | 602 | |||
NM_019007.3 | NP_061880.2 | 3 | 514 | |||
XM_011530976.2 | XP_011529278.1 | 4 | 568 | |||
GenBank mRNA | AK000818.1 | - | 1 | 96 | ||
AK097018.1 | - | 1 | 906 | |||
BC007677.2 | - | 3 | 514 | 3609980 | ||
L20773.1 | - | 1 | 236 |