Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318512.1 | NP_001305441.1 | 2 | 355 | ||
NM_001318514.1 | NP_001305443.1 | 3 | 515 | |||
NM_019009.3 | NP_061882.2 | 3 | 505 | |||
GenBank mRNA | AB209541.1 | - | 3 | 446 | ||
AJ242972.1 | - | 3 | 451 | |||
AK022871.1 | - | 3 | 412 | |||
AK090850.1 | - | 2 | 316 | |||
AK127123.1 | - | 3 | 468 | |||
AK315592.1 | - | 3 | 466 | |||
AL136835.1 | - | 3 | 452 | |||
AY730683.1 | - | 3 | 341 | |||
BC004420.1 | - | 3 | 465 | 3356339 | ||
BC012057.1 | - | 3 | 430 | 4647828 | ||
BC018272.1 | - | 3 | 422 | 3452585 | ||
CR533477.1 | - | 3 | 341 | |||
DB039994.1 | - | 3 | 506 |