Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019083.2 | NP_061956.2 | 11 | 1311 | ||
NR_135078.1 | 11 | 1070 | ||||
XM_005270945.3 | XP_005271002.1 | 11 | 1339 | |||
XM_005270946.2 | XP_005271003.1 | 10 | 1231 | |||
XM_017001504.1 | XP_016856993.1 | 10 | 2963 | |||
XM_017001505.1 | XP_016856994.1 | 10 | 2952 | |||
GenBank mRNA | AK002081.1 | - | 11 | 1298 | ||
BC057226.1 | - | 10 | 878 | |||
BC075811.1 | - | 11 | 1308 | 5104751 |
Gene Symbol: LRRC39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256385.1 | NP_001243314.1 | 11 | 1724 | ||
NM_001256386.1 | NP_001243315.1 | 10 | 1654 | |||
NM_001256387.1 | NP_001243316.1 | 9 | 1597 | |||
NM_144620.3 | NP_653221.1 | 10 | 1637 | |||
GenBank mRNA | N70864.1 | - | 1 | 225 |