Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271562.1 | NP_001258491.1 | 1 | 364 | ||
NM_019021.3 | NP_061894.2 | 1 | 364 | |||
GenBank mRNA | AB101209.1 | - | 1 | 353 | ||
AK000017.1 | - | 1 | 325 | |||
BC009041.1 | - | 1 | 293 | 3868744 | ||
BC071695.1 | - | 1 | 231 | 5416335 | ||
CR457222.1 | - | 1 | 145 | |||
HY013971.1 | - | 1 | 365 |