Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019022.3 | NP_061895.3 | 1, 2 | 183 | ||
XM_005266714.2 | XP_005266771.1 | 1, 2 | 174 | |||
XM_011526033.2 | XP_011524335.1 | 1, 2 | 173 | |||
XM_011526034.2 | XP_011524336.1 | 1, 2 | 175 | |||
XM_011526035.2 | XP_011524337.1 | 1, 2 | 175 | |||
GenBank mRNA | AB058733.1 | - | 1, 2 | 145 | ||
AK122715.1 | - | 1, 2 | 175 | |||
AK295551.1 | - | 1, 2 | 124 | |||
AK307269.1 | - | 1, 2 | 102 | |||
AK310389.1 | - | 1, 2 | 365 | |||
AL832341.1 | - | 1, 2 | 131 | |||
AL832356.1 | - | 1, 2 | 131 | |||
BC032325.2 | - | 1, 2 | 136 | |||
BC093792.1 | - | 1, 2 | 143 | 7939637 | ||
BC093794.1 | - | 1, 2 | 144 | 7939639 | ||
BC107422.1 | - | 1, 2 | 179 | |||
BC143400.1 | - | 1, 2 | 144 | |||
BX647846.1 | - | 1, 2 | 192 |