Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEATR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019024.2 | NP_061897.1 | 20 | 3106 | ||
XM_006712034.2 | XP_006712097.1 | 20 | 3045 | |||
XM_006712035.3 | XP_006712098.1 | 20 | 3109 | |||
XM_011532934.2 | XP_011531236.1 | 20 | 3573 | |||
XM_011532935.2 | XP_011531237.1 | 9 | 1245 | |||
XM_017004378.1 | XP_016859867.1 | 20 | 3045 | |||
XM_017004379.1 | XP_016859868.1 | 21 | 3483 | |||
XM_017004380.1 | XP_016859869.1 | 21 | 3256 | |||
GenBank mRNA | AB037835.1 | - | 11 | 1553 | ||
AK096637.1 | - | 20 | 3045 | |||
BC150605.1 | - | 20 | 3091 | 9056975 | ||
BX538008.1 | - | 14 | 2028 | |||
BX648805.1 | - | 6 | 1266 |