Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184965.1 | NP_001171894.1 | 9 | 1773 | ||
NM_001184966.1 | NP_001171895.1 | 8 | 1698 | |||
NM_019045.4 | NP_061918.3 | 9 | 1773 | |||
XM_011531353.2 | XP_011529655.1 | 8 | 1774 | |||
XM_017029600.1 | XP_016885089.1 | 8 | 1706 | |||
GenBank mRNA | AK001978.1 | - | 3 | 241 | ||
AK092077.1 | - | 6 | 777 | |||
AK299711.1 | - | 8 | 1634 | |||
AK304432.1 | - | 9 | 1300 | |||
AL833605.1 | - | 9 | 1738 | |||
AL834210.1 | - | 6 | 1040 | |||
BC028697.2 | - | 9 | 1709 | 4837674 | ||
BX537578.1 | - | 9 | 1748 | |||
BX538087.1 | - | 9 | 1715 | |||
BX648150.1 | - | 9 | 1712 |