Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 23 | 2578 | ||
NR_125340.1 | 20 | 2325 | ||||
XM_005244931.2 | XP_005244988.1 | 22 | 2518 | |||
XM_006711192.2 | XP_006711255.1 | 24 | 2488 | |||
XM_006711193.3 | XP_006711256.1 | 24 | 2620 | |||
XM_006711194.3 | XP_006711257.1 | 11 | 12616 | |||
XM_011509246.2 | XP_011507548.1 | 23 | 2439 | |||
XM_011509247.1 | XP_011507549.1 | 22 | 2379 | |||
XM_011509248.2 | XP_011507550.1 | 23 | 2372 | |||
XM_011509249.2 | XP_011507551.1 | 24 | 2677 | |||
XM_017000470.1 | XP_016855959.1 | 24 | 2566 | |||
XM_017000471.1 | XP_016855960.1 | 23 | 2428 | |||
XM_017000472.1 | XP_016855961.1 | 19 | 2052 | |||
GenBank mRNA | AK000061.1 | - | 9 | 1115 | ||
AK024832.1 | - | 2 | 462 | |||
AK091207.1 | - | 20 | 2139 | |||
BC074735.2 | - | 6 | 964 |