Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM193B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024019.3 | 8 | 2971 | |||
XM_006714875.2 | XP_006714938.1 | 6 | 1440 | |||
XM_006714876.1 | XP_006714939.1 | 6 | 2163 | |||
XM_006714878.1 | XP_006714941.1 | 7 | 2017 | |||
XM_006714879.3 | XP_006714942.1 | 7 | 1260 | |||
XM_006714880.1 | XP_006714943.1 | 6 | 2058 | |||
XM_006714884.1 | XP_006714947.1 | 4 | 561 | |||
XM_006714885.1 | XP_006714948.1 | 3 | 365 | |||
XM_006714886.2 | XP_006714949.1 | 7 | 1485 | |||
XM_011534572.1 | XP_011532874.1 | 6 | 1335 | |||
XM_011534573.1 | XP_011532875.1 | 7 | 1435 | |||
XM_011534574.1 | XP_011532876.1 | 7 | 1880 | |||
XM_011534579.2 | XP_011532881.1 | 6 | 1199 | |||
XM_011534580.1 | XP_011532882.1 | 7 | 2224 | |||
XM_011534581.1 | XP_011532883.1 | 6 | 1904 | |||
XM_011534582.1 | XP_011532884.1 | 7 | 2411 | |||
XM_011534583.1 | XP_011532885.1 | 7 | 2152 | |||
GenBank mRNA | AB067518.1 | - | 8 | 2928 | ||
AK001266.1 | - | 2 | 241 | |||
AK096032.1 | - | 6 | 1123 | |||
BC008784.2 | - | 4 | 561 | 3626782 | ||
BC086859.1 | - | 4 | 785 | |||
BX647587.1 | - | 5 | 1056 |