Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144952.1 | NP_001138424.1 | 28, 29 | 3990 | ||
XM_011524914.2 | XP_011523216.1 | 28, 29 | 4374 | |||
XM_011524915.2 | XP_011523217.1 | 28, 29 | 4376 | |||
GenBank mRNA | AB040947.1 | - | 21, 22 | 3021 | ||
AL137357.1 | - | 6, 7 | 898 | |||
BC045763.1 | - | 10, 11 | 1518 | |||
BC066363.1 | - | 20, 21 | 2890 |