Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019074.3 | NP_061947.1 | 2 | 510 | ||
GenBank mRNA | AB036931.1 | - | 2 | 450 | ||
AB043894.1 | - | 2 | 174 | |||
AF253468.1 | - | 2 | 174 | |||
AF279305.1 | - | 1 | 80 | |||
AK308755.1 | - | 2 | 503 | |||
AK313831.1 | - | 2 | 505 | |||
AY358894.1 | - | 2 | 209 | |||
BC106950.2 | - | 2 | 310 | 40034887 | ||
DA089757.1 | - | 2 | 510 |