Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276378.1 | NP_001263307.1 | 8 | 950 | ||
NM_001276379.1 | NP_001263308.1 | 7 | 825 | |||
NM_020347.3 | NP_065080.1 | 6 | 669 | |||
NR_075080.1 | 5 | 544 | ||||
XM_006713207.2 | XP_006713270.2 | 6 | 654 | |||
XM_011533838.2 | XP_011532140.1 | 6 | 593 | |||
XM_017006644.1 | XP_016862133.1 | 9 | 1307 | |||
XM_017006645.1 | XP_016862134.1 | 8 | 1182 | |||
GenBank mRNA | AJ297351.1 | - | 6 | 626 | ||
AK093705.1 | - | 8 | 1268 | |||
AK303416.1 | - | 9 | 721 | |||
BC025988.2 | - | 6 | 598 | 4838035 | ||
BC042483.1 | - | 6 | 629 | 4831113 | ||
BX640604.1 | - | 6 | 654 | |||
CN429018.1 | - | 5 | 454 | |||
DA408809.1 | - | 5 | 515 |