Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161407.1 | NP_001154879.1 | 2 | 531 | ||
NM_019080.2 | NP_061953.2 | 2 | 531 | |||
XM_006719840.3 | XP_006719903.1 | 2 | 930 | |||
GenBank mRNA | AB032991.1 | - | 2 | 295 | ||
AB097019.1 | - | 2 | 503 | |||
AB097028.1 | - | 2 | 503 | |||
AB097029.1 | - | 2 | 503 | |||
AB097030.1 | - | 2 | 181 | |||
AB097031.1 | - | 2 | 181 | |||
AK001723.1 | - | 2 | 322 | |||
AK098708.1 | - | 3 | 284 | |||
AK294839.1 | - | 2 | 202 | |||
BC021988.1 | - | 2 | 263 | 4101140 | ||
BC026126.2 | - | 2 | 292 | 3027907 | ||
DC320871.1 | - | 2 | 537 |