Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099658.1 | NP_001093128.1 | 3 | 2563 | ||
NM_001099660.1 | NP_001093130.1 | 4 | 2670 | |||
NM_018334.4 | NP_060804.3 | 2 | 2481 | |||
GenBank mRNA | AB060967.1 | - | 1 | 1930 | ||
AF134481.1 | - | 1 | 1961 | |||
AK292556.1 | - | 2 | 2070 | |||
AL442092.1 | - | 1 | 1886 | |||
AY358280.1 | - | 3 | 2103 | |||
BC035133.2 | - | 2 | 2360 | 5263900 |