Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161779.1 | NP_001155251.1 | 3 | 1822 | ||
NM_001161780.1 | NP_001155252.1 | 3 | 1696 | |||
NM_001161781.1 | NP_001155253.1 | 2 | 1683 | |||
NM_018444.3 | NP_060914.2 | 2 | 1740 | |||
XM_011517135.2 | XP_011515437.1 | 3 | 1593 | |||
XM_011517136.1 | XP_011515438.1 | 3 | 1700 | |||
XM_017013587.1 | XP_016869076.1 | 2 | 1755 | |||
XM_017013588.1 | XP_016869077.1 | 3 | 1663 | |||
GenBank mRNA | AF155661.1 | - | 3 | 1710 | ||
AK022057.1 | - | 1 | 327 | |||
AK057079.1 | - | 2 | 1652 | |||
AK126862.1 | - | 1 | 2117 | |||
AK302651.1 | - | 2 | 1626 | |||
BC047619.1 | - | 2 | 1661 | 4824168 | ||
BC064978.1 | - | 3 | 1680 | |||
BC098343.1 | - | 1 | 1494 | 40010016 |