Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019098.4 | NP_061971.3 | 16 | 1952 | ||
XM_011517138.2 | XP_011515440.1 | 14 | 1607 | |||
GenBank mRNA | AF228520.1 | - | 13 | 1550 | ||
AF272900.1 | - | 16 | 1950 | |||
BC150601.1 | - | 16 | 1930 | 9056971 |