Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020208.3 | NP_064593.1 | 8 | 1383 | ||
NM_022405.3 | NP_071800.1 | 7 | 1272 | |||
XM_011533847.2 | XP_011532149.1 | 7 | 1697 | |||
GenBank mRNA | AF125107.1 | - | 9 | 1447 | ||
AJ276207.1 | - | 8 | 1310 | |||
AJ276208.1 | - | 7 | 1199 | |||
AL389979.1 | - | 3 | 1447 | |||
BC126197.1 | - | 8 | 1323 | 8991913 | ||
BC136431.1 | - | 8 | 1323 | 9020420 | ||
BC144087.1 | - | 8 | 1323 | 9052607 |