Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017515.4 | NP_059985.2 | 5 | 1045 | ||
GenBank mRNA | AK023080.1 | - | 5 | 642 | ||
AK128062.1 | - | 8 | 1214 | |||
AK300582.1 | - | 4 | 480 | |||
AL833969.1 | - | 4 | 1849 | |||
BC039195.2 | - | 5 | 785 | 3847013 | ||
BC048302.1 | - | 5 | 651 | 5499821 |