Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017520.3 | NP_059990.2 | 9, 10 | 2115 | ||
XM_011535124.2 | XP_011533426.1 | 9, 10 | 2116 | |||
GenBank mRNA | AJ293409.1 | - | 4, 5 | 418 | ||
AK056785.1 | - | 2, 3 | 162 | |||
AK300258.1 | - | 9, 10 | 2115 | |||
AL832864.1 | - | 9, 10 | 2064 | |||
BC003542.1 | - | 9, 10 | 2023 | 3611719 | ||
BC046214.1 | - | 3, 4 | 302 |