Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017520.3 | NP_059990.2 | 1 | 199 | ||
XM_011535124.2 | XP_011533426.1 | 1 | 200 | |||
GenBank mRNA | AK092556.1 | - | 1 | 159 | ||
AK300258.1 | - | 1 | 199 | |||
AL832864.1 | - | 1 | 148 | |||
BC003542.1 | - | 1 | 107 | 3611719 | ||
DA672302.1 | - | 1 | 199 |