Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198681.1 | NP_001185610.1 | 4 | 342 | ||
NM_017526.4 | NP_059996.1 | 3 | 478 | |||
GenBank mRNA | AK074841.1 | - | 3 | 239 | ||
AK309370.1 | - | 4 | 342 | |||
BC011027.2 | - | 2 | 186 | |||
BC056250.1 | - | 3 | 223 | 4328021 | ||
CB140261.1 | - | 3 | 346 | |||
CR541647.1 | - | 3 | 201 | |||
CR541737.1 | - | 3 | 201 | |||
Y12670.1 | - | 3 | 271 |
Gene Symbol: LEPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX475249.1 | - | 3 | 291 |