Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318864.1 | NP_001305793.1 | 12 | 1203 | ||
NM_017563.4 | NP_060033.3 | 11 | 1194 | |||
XM_005265238.4 | XP_005265295.1 | 11 | 1405 | |||
XM_006713209.3 | XP_006713272.1 | 13 | 1332 | |||
XM_011533849.2 | XP_011532151.2 | 11 | 989 | |||
GenBank mRNA | AF458067.1 | - | 11 | 1081 | ||
AF494208.1 | - | 11 | 1170 | |||
AF494211.1 | - | 12 | 1171 | |||
AK093074.1 | - | 12 | 1170 | |||
AK302045.1 | - | 11 | 1020 | |||
AL133097.1 | - | 7 | 556 | |||
AL833913.1 | - | 11 | 994 | |||
AY358774.1 | - | 11 | 1117 | |||
AY489047.1 | - | 12 | 1196 | |||
BC038369.1 | - | 12 | 1192 | |||
BC111702.1 | - | 11 | 1021 | 40038598 |