Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017621.3 | NP_060091.1 | 3 | 385 | ||
XM_005250464.2 | XP_005250521.1 | 3 | 278 | |||
XM_005250465.2 | XP_005250522.1 | 3 | 364 | |||
GenBank mRNA | AK000020.1 | - | 3 | 372 | ||
AK026097.1 | - | 3 | 283 | |||
AK222689.1 | - | 3 | 374 | |||
BC002820.2 | - | 3 | 352 | 3636984 | ||
BC017096.1 | - | 3 | 352 | 2820440 | ||
BX353099.2 | - | 3 | 386 |